Learn More
Medchemexpress LLC Rilmenidine phosphate | 85409-38-7 | 99.5% | 278.24 g·mol⁻¹ | C10H19N2O5P | 100 MG
SDP
Catalog No. 5000214956
Encompass_Preferred
Rilmenidine phosphate is the phosphate salt of rilmenidine, an orally active antihypertensive agent that functions as a selective imidazoline and adrenergic receptor agonist. It is supplied for research and analytical applications and has reported cellular activity including modulation of autophagy and induction of apoptosis in select cell models.
  • Orally active antihypertensive agent.
  • Selective I1 imidazoline receptor agonist.
  • Acts as an α2-adrenoceptor agonist.
  • Used as a research and analytical standard.
  • Reported to modulate autophagy and induce apoptosis in cell models.
  • Available in small-scale packages suitable for laboratory use.
Catalog No. 50-002-14956 Supplier Medchemexpress LLC Supplier No. HY100490B100MG
Rilmenidine phosphate is the phosphate salt of rilmenidine, an orally active antihypertensive agent that functions as a selective imidazoline and adrenergic receptor agonist. It is supplied for research and analytical applications and has reported cellular activity including modulation of autophagy and induction of apoptosis in select cell models.
  • Orally active antihypertensive agent.
  • Selective I1 imidazoline receptor agonist.
  • Acts as an α2-adrenoceptor agonist.
  • Used as a research and analytical standard.
  • Reported to modulate autophagy and induce apoptosis in cell models.
  • Available in small-scale packages suitable for laboratory use.