Learn More
Medchemexpress LLC Rilmenidine (phosphate) | 85409-38-7 | MFCD00875846 | 99.5% | 278.24 g/mol | C10H19N2O5P | 50 MG
SDP
Catalog No. 5000214714
Encompass_Preferred
Rilmenidine phosphate is a research-grade antihypertensive compound and selective I1 imidazoline receptor agonist supplied as the phosphate salt for preclinical and in vitro pharmacology studies. It is commonly used to study imidazoline receptor signaling and blood pressure regulation. CAS 85409-38-7; molecular formula C10H19N2O5P; molecular weight 278.24 g/mol; reported purity ~99.48%.
  • Selective I1 imidazoline receptor agonist activity.
  • Provided as the phosphate salt for stability and handling.
  • High purity suitable for research and preclinical assays.
  • Available in small milligram packaging for laboratory use.
  • Supplied with safety data sheet and batch purity documentation.
Catalog No. 50-002-14714 Supplier Medchemexpress LLC Supplier No. HY100490B50MG
Rilmenidine phosphate is a research-grade antihypertensive compound and selective I1 imidazoline receptor agonist supplied as the phosphate salt for preclinical and in vitro pharmacology studies. It is commonly used to study imidazoline receptor signaling and blood pressure regulation. CAS 85409-38-7; molecular formula C10H19N2O5P; molecular weight 278.24 g/mol; reported purity ~99.48%.
  • Selective I1 imidazoline receptor agonist activity.
  • Provided as the phosphate salt for stability and handling.
  • High purity suitable for research and preclinical assays.
  • Available in small milligram packaging for laboratory use.
  • Supplied with safety data sheet and batch purity documentation.