Learn More
Aviva Systems Biology Corp RILP Antibody - C-terminal region (ARP64148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RILP. It was validated on Western Blot. . Alternate names: FLJ31193, PP10141ApplicationWB. Species reactivity: Human
Catalog No. 50-288-188