Learn More
Medchemexpress LLC HY-10574 10mg , Rilpivirine CAS:500287-72-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1057410MG
Encompass_Preferred
HY-10574 10mg Rilpivirine CAS: 500287-72-9 Rilpivirine (R278474) is a potent and specific diarylpyrimidine (DAPY) non-nucleoside reverse transcriptase inhibitor (NNRTI). Rilpivirine has high antiviral activity against wild-type HIV (EC50=0.4 nM) and mutant viruses (EC50=0.1-2.0 nM). Rilpivirine has a high genetic barrier to resistance development of HIV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9445