Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rilpivirine (hydrochloride) | 700361-47-3 | MFCD11046523 | 99.8% | 402.88 | C22H19ClN6 | 100 MG
SDP

Catalog No. 5000240226
Encompass_Preferred

Rilpivirine hydrochloride is the hydrochloride salt of rilpivirine, a diarylpyrimidine non-nucleoside reverse transcriptase inhibitor (NNRTI) used in antiviral research. It is supplied as a solid research reagent suitable for in vitro studies and formulation work.

  • High purity (99.8%).
  • Chemical formula C22H19ClN6; molecular weight 402.88.
  • Solid form, suitable for weighing and dissolution into common solvents.
  • Recommended storage: 4°C sealed; in solution: -80°C (6 months) or -20°C (1 month).
  • Available in 100 mg research packs for laboratory use.

Catalog No. 50-002-40226 Supplier Medchemexpress LLC Supplier No. HY10574A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rilpivirine hydrochloride is the hydrochloride salt of rilpivirine, a diarylpyrimidine non-nucleoside reverse transcriptase inhibitor (NNRTI) used in antiviral research. It is supplied as a solid research reagent suitable for in vitro studies and formulation work.

  • High purity (99.8%).
  • Chemical formula C22H19ClN6; molecular weight 402.88.
  • Solid form, suitable for weighing and dissolution into common solvents.
  • Recommended storage: 4°C sealed; in solution: -80°C (6 months) or -20°C (1 month).
  • Available in 100 mg research packs for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.