Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rilpivirine hydrochloride | 700361-47-3 | MFCD11046523 | 99.8% | 402.88 g·mol-1 | C22H19ClN6 | 1 ML
SDP

Catalog No. 5000240237
Encompass_Preferred

Rilpivirine hydrochloride is the hydrochloride salt of rilpivirine, a diarylpyrimidine non-nucleoside reverse transcriptase inhibitor (NNRTI) used in antiviral research. Supplied as a high-purity solid and as a 10 mM solution in DMSO, it supports biochemical and cell-based assays of HIV reverse transcriptase.

  • High purity suitable for research (99.8%).
  • Available as solid and 10 mM solution in DMSO.
  • Molecular formula C22H19ClN6 and molar mass ~402.88 g·mol-1.
  • Suitable for biochemical and cell-based antiviral assays.
  • Provided with COA and SDS documentation.

Catalog No. 50-002-40237 Supplier Medchemexpress LLC Supplier No. HY10574A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rilpivirine hydrochloride is the hydrochloride salt of rilpivirine, a diarylpyrimidine non-nucleoside reverse transcriptase inhibitor (NNRTI) used in antiviral research. Supplied as a high-purity solid and as a 10 mM solution in DMSO, it supports biochemical and cell-based assays of HIV reverse transcriptase.

  • High purity suitable for research (99.8%).
  • Available as solid and 10 mM solution in DMSO.
  • Molecular formula C22H19ClN6 and molar mass ~402.88 g·mol-1.
  • Suitable for biochemical and cell-based antiviral assays.
  • Provided with COA and SDS documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.