Learn More
Apexbio Technology LLC Riluzole, 100mg. Cas: 1744-22-5 MFCD: MFCD00210213. Sodium channel protein inhibitor
SDP
Supplier:  Apexbio Technology LLC A8513100
Encompass_Preferred
Riluzole, a novel glutamate-modulating agent, has neuroprotective effects [1] [2], via a complex mechanism involving inhibition of voltage-dependent Na channels, high-voltage activated Ca and K channels, and inhibition of protein kinase C. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5156