Learn More
Medchemexpress LLC HY-B0211 100mg , Riluzole CAS:1744-22-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0211100MG
Encompass_Preferred
HY-B0211 100mg Riluzole CAS: 1744-22-5 Riluzole is an anticonvulsant drug and belongs to the family of use-dependent Na+ channel blocker which can also inhibit GABA uptake with an IC50 of 43 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1121