Learn More
Medchemexpress LLC Riluzole-13C,15N2 | 1215552-03-6 | 99.9% | 1 MG
SDP
Catalog No. 5000405518
Encompass_Preferred
Riluzole-13C,15N2 is the 13C and 15N labeled Riluzole. It is an anticonvulsant agent and belongs to the family of use-dependent Na+ channel blockers, inhibiting GABA uptake with an IC50 of 43 μM. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05518 Supplier Medchemexpress LLC Supplier No. HYB0211S1MG
Riluzole-13C,15N2 is the 13C and 15N labeled Riluzole. It is an anticonvulsant agent and belongs to the family of use-dependent Na+ channel blockers, inhibiting GABA uptake with an IC50 of 43 μM. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS