Learn More
Enzo Life Sciences Riluzole (20mg). CAS: 1744-22-5
SDP
Supplier:  Enzo Life Sciences BMLNA1380020
Encompass_Preferred
Glutamate release inhibitor. Neuroprotective. Blocks the inactivated state of voltage-gated neuronal Na+ channels at very low concentrations (about 0.1µM) and activates the TREK family of two-pore domain potassium channels. Alternative name: 2-Amino-6-(trifluoromethoxy)-benzothiazole, 6-(Trifluoromethoxy)-2-benzothiazolamine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml), 100% ethanol (>25mg/ml), methanol (>40mg/ml), 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (14mg/ml) or in 0.1N HCl (11mg/ml); insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-1182