Learn More
Medchemexpress LLC HY-B0211A 50mg Medchemexpress, Riluzole hydrochloride CAS:850608-87-6 Purity:>98%
SDP
Catalog No. 502082410
Encompass_Preferred
Medchemexpress, HY-B0211A 50mg Riluzole hydrochloride CAS:850608-87-6 Riluzole hydrochloride is an anticonvulsant drug and belongs to the family of use-dependent Na+ channel blocker which can also inhibit GABA uptake with an IC50 of 43 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2410 Supplier Medchemexpress LLC Supplier No. HYB0211A50MG
Medchemexpress, HY-B0211A 50mg Riluzole hydrochloride CAS:850608-87-6 Riluzole hydrochloride is an anticonvulsant drug and belongs to the family of use-dependent Na+ channel blocker which can also inhibit GABA uptake with an IC50 of 43 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.