Learn More
Medchemexpress LLC Riluzole hydrochloride | 850608-87-6 | 99.9% | 10 MM * 1 ML
SDP
Catalog No. 5000405715
Encompass_Preferred
Riluzole hydrochloride is an anticonvulsant agent that functions as a use-dependent Na+ channel blocker. It can also inhibit GABA uptake with an IC50 of 43 μM. This product is intended for research use only.
  • Anticonvulsant agent
  • Functions as a use-dependent Na+ channel blocker
  • Inhibits GABA uptake
Catalog No. 50-004-05715 Supplier Medchemexpress LLC Supplier No. HYB0211A10MM/1ML
Riluzole hydrochloride is an anticonvulsant agent that functions as a use-dependent Na+ channel blocker. It can also inhibit GABA uptake with an IC50 of 43 μM. This product is intended for research use only.
  • Anticonvulsant agent
  • Functions as a use-dependent Na+ channel blocker
  • Inhibits GABA uptake