Learn More
Cayman Chemical Riluzole (hydrochloride), Purity: ≥98%, CAS: 850608-87-6, 50 mg
SDP
Catalog No. 502973319
Encompass_Preferred
A benzothiazole derivative with anti-excitotoxic effects that acts by blocking the presynaptic release of glutamate, indirectly antagonizing glutamate receptors, and inactivating neuronal voltage-gated Na+ channels (ED50 = 2.3 μM); suppresses glutamate-induced seizures in rats (ED50 = 3.2 mg/kg) and displays neuroprotective effects in hypoxic animals (ED50 = 4 mg/kg)
Catalog No. 50-297-3319 Supplier Cayman Chemical Supplier No. 1457750 MG
A benzothiazole derivative with anti-excitotoxic effects that acts by blocking the presynaptic release of glutamate, indirectly antagonizing glutamate receptors, and inactivating neuronal voltage-gated Na+ channels (ED50 = 2.3 μM); suppresses glutamate-induced seizures in rats (ED50 = 3.2 mg/kg) and displays neuroprotective effects in hypoxic animals (ED50 = 4 mg/kg)