Learn More
Medchemexpress LLC HY-100779 100mg Medchemexpress, Rimacalib CAS:215174-50-8 Purity:>98%
SDP
Catalog No. 501931946
Encompass_Preferred
Medchemexpress, HY-100779 100mg Rimacalib CAS:215174-50-8 Rimacalib is a Ca 2+ /calmodulin-dependent protein kinase II (CaMKII) inhibitor, with IC 50 s of ~1 μM for CaMKIIα to ~30 μM for CaMKIIγ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1946 Supplier Medchemexpress LLC Supplier No. HY100779100MG
Medchemexpress, HY-100779 100mg Rimacalib CAS:215174-50-8 Rimacalib is a Ca 2+ /calmodulin-dependent protein kinase II (CaMKII) inhibitor, with IC 50 s of ~1 μM for CaMKIIα to ~30 μM for CaMKIIγ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.