Learn More
Med Vet International Zoetis, Rimadyl, 100mg, 30 Caplets
Catalog No. 502827996
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Rimadyl caplets and chewable tablets contain carprofen, an NSAID of the propionic acid class. It is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. For oral use in dogs only.
Catalog No. 50-282-7996 Supplier Med Vet International Supplier No. RXRIM10030CAP
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Rimadyl caplets and chewable tablets contain carprofen, an NSAID of the propionic acid class. It is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. For oral use in dogs only.