Learn More
Bio Serv Rodent MD's™, Rimadyl (2 mg/Tablet), Bacon Flavor, 100/BottleGamma Irradiated
SDP
Catalog No. NC2890078
Encompass
5 gm Tablet - 1" dia. x 1/4" highShelf Life: 6 months, cool dry storageRimadyl (Carprofen) is an effective anti-inflammatory drug used in a variety of species including rodents. Rimadyl MD's are 5 gram size, bacon flavored tablets that meet both the nutritional and pain medication needs of rodents. The highly palatable tablets encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and laborious post-operative injections. Rimadyl MD’s can be customized to include other medications to deliver both medications easily and affordably.
Catalog No. NC2890078 Supplier Bio Serv Supplier No. SMD1502
5 gm Tablet - 1" dia. x 1/4" highShelf Life: 6 months, cool dry storageRimadyl (Carprofen) is an effective anti-inflammatory drug used in a variety of species including rodents. Rimadyl MD's are 5 gram size, bacon flavored tablets that meet both the nutritional and pain medication needs of rodents. The highly palatable tablets encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and laborious post-operative injections. Rimadyl MD’s can be customized to include other medications to deliver both medications easily and affordably.