Learn More
Med Vet International Zoetis, Rimadyl (Carprofen) Injection 50mg/mL 20mL
Catalog No. 502828028
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
The injectable formulation of Rimadyl (carprofen 50 mg/mL) is for SQ use in dogs only. Carprofen, an NSAID of the propionic acid class, is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges*
Catalog No. 50-282-8028 Supplier Med Vet International Supplier No. RXRIMINJ
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
The injectable formulation of Rimadyl (carprofen 50 mg/mL) is for SQ use in dogs only. Carprofen, an NSAID of the propionic acid class, is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges*