Learn More
Apexbio Technology LLC Rimantadine 13392-28-4 200mg
SDP
Catalog No. 5000090083
Encompass_Preferred
Rimantadine (13392-28-4) is a small-molecule inhibitor targeting the M2 ion channel protein of influenza A virus It is designed to specifically block the M2 channel thereby inhibiting the viral uncoating process and preventing subsequent viral entry into host cells Rimantadine exerts its biological activity primarily through inhibition of the M2 ion channel In in vitro studies rimantadine demonstrates antiviral activity with reported IC50 values typically ranging from approximately 0 1 to 1 M depending on virus strain and experimental context Based on these pharmacological properties rimantadine holds research potential in investigating influenza A viral replication resistance mutations infection pathways and in evaluating antiviral activity and establishing infection models in vitro
Catalog No. 50-000-90083 Supplier Apexbio Technology LLC Supplier No. B2128200
Rimantadine (13392-28-4) is a small-molecule inhibitor targeting the M2 ion channel protein of influenza A virus It is designed to specifically block the M2 channel thereby inhibiting the viral uncoating process and preventing subsequent viral entry into host cells Rimantadine exerts its biological activity primarily through inhibition of the M2 ion channel In in vitro studies rimantadine demonstrates antiviral activity with reported IC50 values typically ranging from approximately 0 1 to 1 M depending on virus strain and experimental context Based on these pharmacological properties rimantadine holds research potential in investigating influenza A viral replication resistance mutations infection pathways and in evaluating antiviral activity and establishing infection models in vitro