Learn More
Apexbio Technology LLC Rimantadine Hydrochloride 1501-84-4 1g
SDP
Catalog No. 5000091211
Encompass_Preferred
Rimantadine Hydrochloride (1501-84-4) is a small-molecule inhibitor targeting influenza viruses It acts by inhibiting viral RNA synthesis thereby reducing influenza A virus replication through interference with viral entry or early replication stages Rimantadine Hydrochloride exerts its biological activity primarily by inhibiting viral replication In cellular assays it demonstrates inhibitory effects on dengue virus serotypes and suppresses viral replication in human peripheral blood leukocytes and rhesus monkey lymphocyte cultures In human lymphocyte cultures at approximately 25 g/ml it inhibits mitogen- and antigen-induced lymphocyte proliferation Based on these pharmacological properties Rimantadine Hydrochloride holds research potential in antiviral studies and cellular immune response assays involving influenza and dengue virus infections
Catalog No. 50-000-91211 Supplier Apexbio Technology LLC Supplier No. B78491000
Add to Cart
Add to Cart
Rimantadine Hydrochloride (1501-84-4) is a small-molecule inhibitor targeting influenza viruses It acts by inhibiting viral RNA synthesis thereby reducing influenza A virus replication through interference with viral entry or early replication stages Rimantadine Hydrochloride exerts its biological activity primarily by inhibiting viral replication In cellular assays it demonstrates inhibitory effects on dengue virus serotypes and suppresses viral replication in human peripheral blood leukocytes and rhesus monkey lymphocyte cultures In human lymphocyte cultures at approximately 25 g/ml it inhibits mitogen- and antigen-induced lymphocyte proliferation Based on these pharmacological properties Rimantadine Hydrochloride holds research potential in antiviral studies and cellular immune response assays involving influenza and dengue virus infections