Learn More
Medchemexpress LLC Rimantadine hydrochloride | 1501-84-4 | 98.0% | 5 G
SDP
Catalog No. 5000407206
Encompass_Preferred
Rimantadine hydrochloride is an orally active inhibitor for M2 protein, designed for research applications. It functions by blocking hydrogen ion channel activity, thereby preventing viral entry and replication, and demonstrating broad-spectrum antiviral effects. This compound is intended for research use only.
  • Orally active M2 protein inhibitor
  • Blocks hydrogen ion channel activity
  • Prevents virus entry and replication
  • Exhibits broad-spectrum antiviral activity
  • Inhibits RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells
  • Induces autophagy in HAV-infected Huh7 cells
  • Exhibits anti-infectious activity in H3N2-infected mouse models
Catalog No. 50-004-07206 Supplier Medchemexpress LLC Supplier No. HYB0338A5G
Rimantadine hydrochloride is an orally active inhibitor for M2 protein, designed for research applications. It functions by blocking hydrogen ion channel activity, thereby preventing viral entry and replication, and demonstrating broad-spectrum antiviral effects. This compound is intended for research use only.
  • Orally active M2 protein inhibitor
  • Blocks hydrogen ion channel activity
  • Prevents virus entry and replication
  • Exhibits broad-spectrum antiviral activity
  • Inhibits RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells
  • Induces autophagy in HAV-infected Huh7 cells
  • Exhibits anti-infectious activity in H3N2-infected mouse models