Learn More
Selleck Chemical LLC Rimantadine Hydrochloride
SDP
Catalog No. 5000003578
Encompass_Preferred
Rimantadine is an RNA synthesis inhibitor that is used as an antiviral agent in the prophylaxis and treatment of influenza It works by inhibiting a viral protein called M2 which normally mediates the transport of acid ions across the influenza virion shell
Catalog No. 50-000-03578 Supplier Selleck Chemical LLC Supplier No. S548425MG
Rimantadine is an RNA synthesis inhibitor that is used as an antiviral agent in the prophylaxis and treatment of influenza It works by inhibiting a viral protein called M2 which normally mediates the transport of acid ions across the influenza virion shell