Learn More
Medchemexpress LLC Rimcazole dihydrochloride | 75859-03-9 | 100.0% | 394.38 | 25 MG
SDP
Catalog No. 5000347246
Encompass_Preferred
Rimcazole dihydrochloride is a carbazole derivative that acts in part as a sigma (σ) receptor antagonist. It also binds with moderate affinity to the dopamine transporter and inhibits dopamine uptake. It can reduce locomotor activity and sensitization and can be used for cancer research.
  • Acts as a sigma (σ) receptor antagonist
  • Binds to the dopamine transporter
  • Inhibits dopamine uptake
  • Reduces locomotor activity and sensitization
  • Can be used for cancer research
Catalog No. 50-003-47246 Supplier Medchemexpress LLC Supplier No. 25MGHY108510
Rimcazole dihydrochloride is a carbazole derivative that acts in part as a sigma (σ) receptor antagonist. It also binds with moderate affinity to the dopamine transporter and inhibits dopamine uptake. It can reduce locomotor activity and sensitization and can be used for cancer research.
  • Acts as a sigma (σ) receptor antagonist
  • Binds to the dopamine transporter
  • Inhibits dopamine uptake
  • Reduces locomotor activity and sensitization
  • Can be used for cancer research