Learn More
Medchemexpress LLC Rimcazole dihydrochloride | 75859-03-9 | 99.96% | 394.38 | 1 MG
SDP
Catalog No. 5000346396
Encompass_Preferred
Rimcazole dihydrochloride (BW 234U) is a carbazole derivative that acts as a sigma (σ) receptor antagonist. It also binds with moderate affinity to the dopamine transporter and inhibits dopamine uptake. It can reduce locomotor activity and sensitization, and is used for cancer research.
  • Acts as a sigma (σ) receptor antagonist.
  • Binds to the dopamine transporter.
  • Inhibits dopamine uptake.
  • Reduces locomotor activity and sensitization.
  • Can be used for cancer research.
  • Induces transient accumulation of HIF-1α protein in cancer cells.
Catalog No. 50-003-46396 Supplier Medchemexpress LLC Supplier No. HY10851010MM/1ML
Rimcazole dihydrochloride (BW 234U) is a carbazole derivative that acts as a sigma (σ) receptor antagonist. It also binds with moderate affinity to the dopamine transporter and inhibits dopamine uptake. It can reduce locomotor activity and sensitization, and is used for cancer research.
  • Acts as a sigma (σ) receptor antagonist.
  • Binds to the dopamine transporter.
  • Inhibits dopamine uptake.
  • Reduces locomotor activity and sensitization.
  • Can be used for cancer research.
  • Induces transient accumulation of HIF-1α protein in cancer cells.