Learn More
Medchemexpress LLC Rimcazole (dihydrochloride) | 75859-03-9 | MFCD00083199 | 100.0% | 394.38 g/mol | C21H29Cl2N3 | 5 MG
SDP
Catalog No. 5000263295
Encompass_Preferred
Rimcazole dihydrochloride is a carbazole-derivative research compound that functions as a sigma (σ) receptor antagonist and interacts with the dopamine transporter. It is supplied as the dihydrochloride salt (CAS 75859-03-9) with molecular formula C21H29Cl2N3 and molecular weight 394.38 g/mol.
  • Acts as a sigma (σ) receptor antagonist.
  • Interacts with dopamine transporter and inhibits dopamine uptake.
  • Provided as the dihydrochloride salt (CAS 75859-03-9).
  • Supplied in small mass quantities suitable for laboratory research, e.g., 5 mg.
  • High reported purity suitable for analytical and pharmacological studies.
Catalog No. 50-002-63295 Supplier Medchemexpress LLC Supplier No. HY1085105MG
Rimcazole dihydrochloride is a carbazole-derivative research compound that functions as a sigma (σ) receptor antagonist and interacts with the dopamine transporter. It is supplied as the dihydrochloride salt (CAS 75859-03-9) with molecular formula C21H29Cl2N3 and molecular weight 394.38 g/mol.
  • Acts as a sigma (σ) receptor antagonist.
  • Interacts with dopamine transporter and inhibits dopamine uptake.
  • Provided as the dihydrochloride salt (CAS 75859-03-9).
  • Supplied in small mass quantities suitable for laboratory research, e.g., 5 mg.
  • High reported purity suitable for analytical and pharmacological studies.