Learn More
Selleck Chemical LLC Rimegepant BMS-927711-5mg
SDP
Supplier:  Selleck Chemical LLC S66595MG
Encompass_Preferred
Rimegepant (BMS-927711, BHV-3000) is a potent, selective, competitive human calcitonin gene-related peptide (CGRP) receptor antagonist with ki of 0.027 nM.
Catalog No. 50-238-9890