Learn More
Selleck Chemical LLC Rimiducid AP1903-5mg
SDP
Catalog No. 502389639
Encompass_Preferred
Rimiducid (AP1903) is a chemical protein dimerizer and binds tightly to the target FKBP fusion protein while binding minimally to the abundant natural FKBP. Rimiducid elicites potent and dose-dependent apoptosis of cells in culture expressing dimerizer-dependent Fas constructs, with an EC50 of ≈0.1 nM.
Catalog No. 50-238-9639 Supplier Selleck Chemical LLC Supplier No. S97265MG
Rimiducid (AP1903) is a chemical protein dimerizer and binds tightly to the target FKBP fusion protein while binding minimally to the abundant natural FKBP. Rimiducid elicites potent and dose-dependent apoptosis of cells in culture expressing dimerizer-dependent Fas constructs, with an EC50 of ≈0.1 nM.