Learn More
MSC Bouton Zenon Safety Readers, Polycarbonate, Clear, +1.50 Diopter, 1 ea
Catalog No. 5000687904
Encompass
These polycarbonate safety glasses feature a +1.50 diopter reading magnification and a lightweight, rimless design suitable for general manufacturing, construction, and indoor work environments. They are engineered for comfort and a secure fit, meeting stringent safety standards.
  • Lightweight, rimless design for all-day user comfort
  • Flexible, rubber-tip temples for a secure fit without pressure points
  • Molded nose bridge designed to fit most users
  • Durable 10 base curve polycarbonate lens with anti-scratch coating
  • Includes UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-006-87904 Supplier MSC Supplier No. 48020994
These polycarbonate safety glasses feature a +1.50 diopter reading magnification and a lightweight, rimless design suitable for general manufacturing, construction, and indoor work environments. They are engineered for comfort and a secure fit, meeting stringent safety standards.
  • Lightweight, rimless design for all-day user comfort
  • Flexible, rubber-tip temples for a secure fit without pressure points
  • Molded nose bridge designed to fit most users
  • Durable 10 base curve polycarbonate lens with anti-scratch coating
  • Includes UV protection
  • Meets ANSI Z87.1+ standards