Learn More
Apexbio Technology LLC Rimonabant, 100mg. Cas: 168273-06-1 MFCD: MFCD04034714. CB1 receptor antagonist
SDP
Supplier:  Apexbio Technology LLC B1429100
Encompass_Preferred
Rimonabant (SR141716) is a potent and selective antagonist of central cannabinoid (CB1) receptor with Ki values of 1.8nM and 514nM for CB1 and CB2, respectively. 168273-06-1. MFCD04034714
Catalog No. 50-101-2409