Learn More
Apexbio Technology LLC Rimonabant hydrochloride 158681-13-1 1g
SDP
Catalog No. 5000096898
Encompass_Preferred
Rimonabant hydrochloride (CAS 158681-13-1) is a small-molecule inverse agonist targeting the cannabinoid receptor subtype CB1 It is designed to modulate endocannabinoid signaling pathways thereby regulating processes related to appetite control lipid metabolism and energy homeostasis Rimonabant hydrochloride exerts its biological activity primarily through selective inverse agonism at CB1 receptors It demonstrates high receptor binding affinity with a Ki value of approximately 1 8 nM Based on these pharmacological properties Rimonabant hydrochloride holds research potential in cannabinoid receptor pharmacology metabolic regulation and obesity-associated disorders
Catalog No. 50-000-96898 Supplier Apexbio Technology LLC Supplier No. A37661000
Add to Cart
Add to Cart
Rimonabant hydrochloride (CAS 158681-13-1) is a small-molecule inverse agonist targeting the cannabinoid receptor subtype CB1 It is designed to modulate endocannabinoid signaling pathways thereby regulating processes related to appetite control lipid metabolism and energy homeostasis Rimonabant hydrochloride exerts its biological activity primarily through selective inverse agonism at CB1 receptors It demonstrates high receptor binding affinity with a Ki value of approximately 1 8 nM Based on these pharmacological properties Rimonabant hydrochloride holds research potential in cannabinoid receptor pharmacology metabolic regulation and obesity-associated disorders