Learn More
TARGETMOL CHEMICALS INC RIMONABANT HYDROCHLORIDE 25MG
SDP
Catalog No. 502577362
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rimonabant hydrochloride (SR 141716A) is a cannabinoid receptor antagonist binding selectively to central cannabinoid receptors (CB1) with high affinity. purity: 99%
Catalog No. 50-257-7362 Supplier TARGETMOL CHEMICALS INC Supplier No. T151925MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rimonabant hydrochloride (SR 141716A) is a cannabinoid receptor antagonist binding selectively to central cannabinoid receptors (CB1) with high affinity. purity: 99%