Learn More
Selleck Chemical LLC Rimonabant hydrochloride E4858-1G
SDP
Catalog No. 5000043286
Encompass_Preferred
Rimonabant hydrochloride (SR 141716A hydrochloride) is a highly potent and selective antagonist of the central cannabinoid receptor (CB1) with a Ki of 1 8 nM It is an anorectic anti-obesity drug with potential to treat obesity and cardiovascular risk factors
Catalog No. 50-000-43286 Supplier Selleck Chemical LLC Supplier No. E48581G
Rimonabant hydrochloride (SR 141716A hydrochloride) is a highly potent and selective antagonist of the central cannabinoid receptor (CB1) with a Ki of 1 8 nM It is an anorectic anti-obesity drug with potential to treat obesity and cardiovascular risk factors