Learn More
Medchemexpress LLC HY-114491 5mg , Rineterkib CAS:1715025-32-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1144915MG
Encompass_Preferred
HY-114491 5mg Rineterkib CAS: 1715025-32-3 Rineterkib (compound B) is an orally available ERK1 and ERK2 inhibitor in the treatment of a proliferative disease characterized by activating mutations in the MAPK pathway. The activity is particularly related to the treatment of KRAS-mutant NSCLC, BRAF-mutant NSCLC, KRAS-mutant pancreatic cancer, KRAS-mutant colorectal cancer (CRC) and KRAS-mutant ovarian cancer. Rineterkib hydrochloride can also inhibit RAF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9637