Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Ring for 2.5mL Syringe AT/KF

Catalog No. 5000207880 Shop All Hach Company Products
Encompass

Ring for 2 5 mL Syringe-AT/KF Titrator

Catalog No. 50-002-07880 Supplier Hach Company Supplier No. LZE104
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ring for 2 5 mL Syringe-AT/KF Titrator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.