Learn More
Chemglass Life Sciences Loosening Ring, Rodaviss, Size 24
SDP
Catalog No. 501531259
Encompass_Preferred
Loosening Ring, Rodaviss, Size 24. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.
Catalog No. 50-153-1259 Supplier Chemglass Life Sciences Supplier No. CG18403
Loosening Ring, Rodaviss, Size 24. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.