Learn More
Hach Company Ring for 25 mL Syringe, AT/KF Titrator
Supplier:  Hach Company LZE107
Encompass
Replacement ring designed to hold or secure 25 mL glass syringes used in AT/KF titrators. It supports the syringe assembly during Karl Fischer and other titration workflows on compatible AT-series instruments.
  • Fits 25 mL syringes.
  • Designed for AT/KF titrators.
  • Supports the syringe assembly during titration.
  • Helps maintain alignment and proper sealing of the syringe.
Catalog No. NC3368695