Learn More
Hach Company Ring for 5 mL Syringe for AT/KF Titrator (Hach LZE105)
Supplier:  Hach Company LZE105
Encompass
Replacement O-ring for 5 mL syringe used with Hach TitraLab AT and KF titration workstations. Restores the syringe seal to maintain accurate volume delivery and prevent leaks during titration procedures.
  • Replacement O-ring for 5 mL syringe.
  • Restores syringe seal to prevent leaks.
  • Helps maintain accurate volumetric delivery during titration.
  • Simple to install for routine syringe maintenance and replacement.
Catalog No. NC3368686