Learn More
Fine Science Tools RING FRCPS
SDP
Catalog No. NC9128323
Encompass
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 6 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 9cm
Catalog No. NC9128323 Supplier Fine Science Tools Supplier No. 1110609
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 6 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 9cm