Learn More
Perkin Elmer US LLC Complete Quartz Torch O-Ring Kit for Optima 2x00/4x00/5x00/7x00 DV
Catalog No. 509043789
Encompass_Preferred
This kit includes: O-Ring 29.74 mm I.D., Qty. 1 (09902131) O-Ring 20.3 mm I.D., Qty. 2 (09902223) O-Ring 28.17 mm I.D., Qty. 1 (09921061) O-Ring 26.57 mm I.D., Qty. 1 (09921062) O-Ring 20.3 mm I.D., Qty. 1 (09902045)
Catalog No. 50-904-3789 Supplier Perkin Elmer US LLC Supplier No. N0770437
This kit includes: O-Ring 29.74 mm I.D., Qty. 1 (09902131) O-Ring 20.3 mm I.D., Qty. 2 (09902223) O-Ring 28.17 mm I.D., Qty. 1 (09921061) O-Ring 26.57 mm I.D., Qty. 1 (09921062) O-Ring 20.3 mm I.D., Qty. 1 (09902045)