Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waldmann Lighting™ RING LED Magnifier Light With 15.7 in. Arm Reach

Catalog No. 11000116
Encompass
Encompass_Preferred
Change view
Click to view available options
Magnification:
2.5x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
11-000-116 2.5x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-000-116 Supplier Waldmann Lighting™ Supplier No. 11314200000618824
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waldmann Lighting™ RING LED magnifier light provides 6 diopters (2.5X) of magnification surrounded by 63 glare-free LEDs.

  • 63 bright glare-free LEDs surround the 6 diopter magnifier lens
  • Gooseneck arm with 15.7 in. reach is strong and flexible, even after 20,000 movements
  • 2.8 in. diameter distortion-free lens specially hard-coated for scratch-resistance
  • Soft-touch sensor on luminaire head switches light on easily
  • Maintenance-free LEDs with over 50,000 hour lamp life

Specifications

Magnification 2.5x
Material Anodized Aluminum
Includes Gooseneck Arm; CE
Diopter 6 diopers
Color Silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.