Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Ring Magnets, Set of 6
GREENER_CHOICESCI_ED

Catalog No. S139439 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139439 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139439 Supplier Supertek™ Supplier No. PH40092S3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ring magnets to use for determining if an object is magnetic.

  • Ring Magnets carry countless uses in the magnetic world.
  • A set of 2 or more ring magnets can be used to demonstrate attraction and repulsion between poles better known as the Floating Magnets Experiment.

Specifications

Grade 1+
Format Kit
Languages English
Subject Physical Science - Magnetism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.