Learn More
Thomson Instrument Company Ring Only for Inverting Optimum Growth Flasks
SDP
Catalog No. NC0530760
Encompass_Preferred
Designed for use with Optimum Growth 5 L Inversion Transfer Caps, this non-sterile ring provides secure support for flask inversion during liquid transfer procedures. Requires a 22" stand for proper use. Ideal for labs looking to replace or separately purchase the ring component of the full stand setup. Made in the USA
Catalog No. NC0530760 Supplier Thomson Instrument Company Supplier No. 931607
Designed for use with Optimum Growth 5 L Inversion Transfer Caps, this non-sterile ring provides secure support for flask inversion during liquid transfer procedures. Requires a 22" stand for proper use. Ideal for labs looking to replace or separately purchase the ring component of the full stand setup. Made in the USA