Learn More
Waters Corp Piston Seal, P-50/FDM, PEEK
Supplier:  Waters Corp 700006757
Encompass_Preferred
This seal is designed for P-50/FDM systems. It is made from PEEK material, ensuring compatibility and performance in critical applications.
  • Designed for P-50/FDM systems
  • Constructed from PEEK material
  • Suitable for SFC system type
  • Comes with one unit per package
  • Replacement is recommended as required
Catalog No. 50-239-1614