Learn More
World Precision Instrument Ring Tipped Forceps, 10cm, Straight, 4.8mm ID
SDP
Catalog No. 502521302
Encompass
WPI Ring-Tipped Forceps with Securing Screw give you a safe and secure means of handling delicate tissues. The screw or locking mechanism allows the forceps to maintain a constant grip on the item being handled. Once the forceps have grasped an object or tissue, tighten the screw to lock the forceps in place, preventing accidental slipping or release. The locking feature of the forceps ensures stability, particularly when the surgeon or researcher needs to maintain a precise position without having to apply continuous manual pressure. By locking the forceps, you can free up your hands for other tasks while ensuring that the forceps maintain their grip. In surgical procedures, like orthopedics applications, the locking forceps can be used to provide a constant and controlled pull on tissues without the need for a continuous manual hold.
Catalog No. 50-252-1302 Supplier World Precision Instrument Supplier No. 503202
WPI Ring-Tipped Forceps with Securing Screw give you a safe and secure means of handling delicate tissues. The screw or locking mechanism allows the forceps to maintain a constant grip on the item being handled. Once the forceps have grasped an object or tissue, tighten the screw to lock the forceps in place, preventing accidental slipping or release. The locking feature of the forceps ensures stability, particularly when the surgeon or researcher needs to maintain a precise position without having to apply continuous manual pressure. By locking the forceps, you can free up your hands for other tasks while ensuring that the forceps maintain their grip. In surgical procedures, like orthopedics applications, the locking forceps can be used to provide a constant and controlled pull on tissues without the need for a continuous manual hold.