Learn More
Aviva Systems Biology Corp RING1 Antibody - N-terminal region (ARP38201_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38201_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RING1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RING1. Alternate names: RNF1, RING1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56562