Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ringer's Solution
SCI_ED

Catalog No. S25513 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Liquid - Amphibian
Liquid - Mammalian
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25513 500 mL Liquid - Mammalian
S25512 500 mL Liquid - Amphibian
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25513
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Ringer's Solution
Color Colorless
CAS 7647-14-5, 7732-18-5
Assay Percent Range >98%
Density 1g/mL
Synonym Physiological Saline
Packaging Liquid - Mammalian
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.