These extrication gloves are designed for emergency professionals, offering critical protection and performance. They feature a waterproof and breathable Hipora® barrier that guards against microorganisms and meets OSHA bloodborne pathogen standards, combined with advanced material construction for enhanced dexterity and safety.
- Equipped with a waterproof and breathable Hipora® barrier against microorganisms
- Meets OSHA Bloodborne Pathogens Standard 29 CFR 1910.1030
- Features 3M™ reflective fabric for visibility
- Boxed fingertip design for improved fit
- Molded TPR panels reduce impact and enhance dexterity
- Elastic cuff and gaiter protect against glass and debris
- Features 0.8mm synthetic leather palm with anti-bacterial properties
- Double Kevlar® stitching in high-wear areas
- Cut-resistant Kevlar® palm, side, thumb, and finger panels