Learn More
MSC Ringers Gloves Work Gloves, Small, Nitrile-Coated, Nylon/Spandex, High-Visibility, 1 Pair
Catalog No. 5000803351
Encompass
Ringers Gloves R840 work gloves are general-purpose gloves with a nitrile-coated palm and high visibility, suitable for diverse applications.
  • Black and lime green; high visibility
  • Nylon and spandex lined back and cuff materials
  • Smooth, palm-dipped nitrile coating for enhanced grip
  • Static dissipative properties
  • Manufactured in Sri Lanka from recycled materials
  • General purpose; ideal for driving, parts picking, handling, packaging, and assembly
  • Small glove size (numeric size 7); full fingered, wing thumb type
  • ANSI/ISEA abrasion resistance level 4
  • Complies with EN 407 X1XXXX, CAT III 0493, CE, ANSI/ISEA Cut Level A1, EN ISO 21420:2020, and EN 388:2016 4131X standards
  • Not FDA approved, not lint-free, not seamless
  • Unisex design; maximum temperature of 104 °F
Catalog No. 50-008-03351 Supplier MSC Supplier No. 70817945
Ringers Gloves R840 work gloves are general-purpose gloves with a nitrile-coated palm and high visibility, suitable for diverse applications.
  • Black and lime green; high visibility
  • Nylon and spandex lined back and cuff materials
  • Smooth, palm-dipped nitrile coating for enhanced grip
  • Static dissipative properties
  • Manufactured in Sri Lanka from recycled materials
  • General purpose; ideal for driving, parts picking, handling, packaging, and assembly
  • Small glove size (numeric size 7); full fingered, wing thumb type
  • ANSI/ISEA abrasion resistance level 4
  • Complies with EN 407 X1XXXX, CAT III 0493, CE, ANSI/ISEA Cut Level A1, EN ISO 21420:2020, and EN 388:2016 4131X standards
  • Not FDA approved, not lint-free, not seamless
  • Unisex design; maximum temperature of 104 °F