Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rink Acid Resin

Catalog No. 5165975G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-659-75G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 51-659-75G Supplier Sigma Aldrich Supplier No. 5165975G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acid labile resin used in Fmoc-based solid phase peptide synthesis (SPPS). Mild acidic cleavage conditions lead to the release of the peptide acid where fully protected peptides can be released if desired. Cleavage of the final protected peptide can be achieved in as little as 10% AcOH in DCM or 1% TFA in DCM.

Specifications

CAS 1303-32-8
Melting Point 98°C to 102°C
Molecular Formula C12H24B2O4
MDL Number MFCD06246008
Quantity 1 g
Molecular Weight (g/mol) 253.94 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.