Learn More
Sigma Aldrich Fine Chemicals Biosciences Rink Amide AM resin 100-20
Catalog No. 502899027
Encompass
This support comprises the modified Rink amide linker attached to aminomethylpolystyrene and is an ideal tool for the fmoc SPPS of peptide amides. Cleavage from this resin can be effected by a single step treatment with 95% TfA providing peptide amides in high yields and purities.
Catalog No. 50-289-9027 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8551300005
This support comprises the modified Rink amide linker attached to aminomethylpolystyrene and is an ideal tool for the fmoc SPPS of peptide amides. Cleavage from this resin can be effected by a single step treatment with 95% TfA providing peptide amides in high yields and purities.