Learn More
Chem-Impex International, Inc. Rink Amide AM Resin, High Yield, High Purity, Efficient Peptide Synthesis
Catalog No. NC3315297
Encompass_Preferred
Rink Amide AM Resin is a high-performance solid-phase support for efficient C-terminal peptide amide synthesis via Fmoc-based Solid-Phase Peptide Synthesis (SPPS). Its unique acid-labile Rink amide linker on a polystyrene-divinylbenzene copolymer ensures mechanical stability and excellent swelling properties in SPPS solvents. Ideal for drug discovery and biochemical studies, it offers high yields and purity of synthesized peptides, setting it apart from alternatives.
Catalog No. NC3315297 Supplier Chem-Impex International, Inc. Supplier No. 06761100G
Rink Amide AM Resin is a high-performance solid-phase support for efficient C-terminal peptide amide synthesis via Fmoc-based Solid-Phase Peptide Synthesis (SPPS). Its unique acid-labile Rink amide linker on a polystyrene-divinylbenzene copolymer ensures mechanical stability and excellent swelling properties in SPPS solvents. Ideal for drug discovery and biochemical studies, it offers high yields and purity of synthesized peptides, setting it apart from alternatives.